Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARD3/Par3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50
Specifications
Antigen | PARD3/Par3 |
---|---|
Concentration | 0.5 mg/ml |
Dilution | Western Blot 1:250 |
Applications | Western Blot |
Classification | Polyclonal |
Description
PARD3/Par3 Polyclonal specifically detects PARD3/Par3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PARD3/Par3 | |
Western Blot 1:250 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog | |
Synthetic peptide from amino acids 31-342 corresponding to rat PAR2 protein from the following sequence, was selected from the unique region of the mouse PAR3 protein, peptide was post-synthetically modified to achieve highest antigenicity before used for coupling to KLH using heterobifunctional cross linker for immunogen preparation. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
0.5 mg/ml | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. | |
56288 | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title