Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556475
Description
Immunogen sequence: LKGLGDMFRI QAKTREFRER QARERDYAEI QDFHRTFGCD DELMYGGVSS YEGSMALNAR PQSPREGHMM DALYAQVKKP RNSKPSPVDS NR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
Specifications
PARD3 | |
Polyclonal | |
Unconjugated | |
Pard3 | |
AA960621; AI256638; ASBP; ASIP; atypical PKC isotype-specific interacting protein; atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; atypical PKC-specific-binding protein; Baz; bazooka; CTCL tumor antigen se2-5; D8Ertd580e; Ephrin-interacting protein; FLJ21015; PAR3; Par-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3A; PAR3alpha; PAR3-alpha; Pard3; PARD-3; PARD3A; partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; PPP1R118; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; SE2-5L16; SE2-5LT1; SE2-5T2; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TEW0 | |
Pard3 | |
Recombinant protein corresponding to Human PARD3. Recombinant protein control fragment (Product #RP-94754). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction