Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572553
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572553 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572553 Supplier Invitrogen™ Supplier No. PA572553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARD3 Polyclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pard3
Gene Accession No. Q8TEW0, Q99NH2
Gene Alias AA960621; AI256638; ASBP; ASIP; atypical PKC isotype-specific interacting protein; atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; atypical PKC-specific-binding protein; Baz; bazooka; CTCL tumor antigen se2-5; D8Ertd580e; Ephrin-interacting protein; FLJ21015; PAR3; Par-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3A; PAR3alpha; PAR3-alpha; Pard3; PARD-3; PARD3A; partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; PPP1R118; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; SE2-5L16; SE2-5LT1; SE2-5T2; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect)
Gene Symbols Pard3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1329-1356 amino acids from the C-terminal region of human PARD3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56288, 93742
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.