Learn More
PARD3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304496AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
PARD proteins, which were first identified in C. elegans, are essential for asymmetric cell division and polarized growth, whereas CDC42 mediates the establishment of cell polarity. The CDC42 GTPase, which is controlled by nucleotide exchange factors and GTPase-activating proteins domain.Specifications
| PARD3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8TEW0, Q99NH2 | |
| PARD3 | |
| Between 1306 and 1356 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PARD3 | |
| RP11-406D17.1, ASIP, Baz, Bazooka, FLJ21015, PAR3, PAR3alpha, PARD3A, SE2-5L16, SE2-5LT1, SE2-5T2, CTCL tumor antigen se2-5; PAR3-alpha; PARD-3; atypical PKC isotype-specific interacting protein; atypical PKC isotype-specific-interacting protein; partitioning defective 3 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56288, 93742 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.