Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARD3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557214 Supplier Invitrogen™ Supplier No. PA557214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARREGFPLYE DDEGRARPSE YDLLWVPGRG PDGNAHNLRF EGMERQYASL PRGGPADPVD YLPAAPRGLY KERELPYYPG AHPMHPPKGS YPRPTELRVA DLRYPQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD3B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pard3b
Gene Accession No. Q8TEW8
Gene Alias 1810008K04Rik; 2010002N16Rik; 2810455B10Rik; Als2cr19; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein homolog; C. elegans; DN-276O13.2; par-3 family cell polarity regulator beta; par-3 partitioning defective 3 homolog B; PAR3B; PAR3beta; PAR3-beta; PAR3L; PAR3-L protein; Pard3b; Partitioning defective 3 homolog B; partitioning defective 3-like protein; partitioning-defective 3-beta; RP23-20G7.1
Gene Symbols Pard3b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARD3B. Recombinant protein control fragment (Product #RP-96042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.