Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARD3B Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537974
Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Specifications
| PARD3B | |
| Polyclonal | |
| Unconjugated | |
| Pard3b | |
| 1810008K04Rik; 2010002N16Rik; 2810455B10Rik; Als2cr19; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein homolog; C. elegans; DN-276O13.2; par-3 family cell polarity regulator beta; par-3 partitioning defective 3 homolog B; PAR3B; PAR3beta; PAR3-beta; PAR3L; PAR3-L protein; Pard3b; Partitioning defective 3 homolog B; partitioning defective 3-like protein; partitioning-defective 3-beta; RP23-20G7.1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 72823 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9CSB4 | |
| Pard3b | |
| Peptide with sequence C-RNQDPRQKNP, from the C-Terminus of mouse Pard3b (aa 1189-1199). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction