Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARD6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101739 Supplier Invitrogen™ Supplier No. PA5101739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PARD6B.

This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain, an OPR domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cytoplasmic protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD6B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pard6b
Gene Accession No. Q9BYG5, Q9JK83
Gene Alias AV025615; par-6 (partitioning defective 6) homolog beta; PAR-6 beta; par-6 family cell polarity regulator beta; par-6 partitioning defective 6 homolog beta; par-6 partitioning defective 6 homolog beta (C. elegans); PAR6B; PAR-6B; Pard6b; pard6beta; partitioning defective 6 beta; partitioning defective 6 homolog beta; partitioning-defective protein 6, beta; si:dkey-192i18.6
Gene Symbols Pard6b
Host Species Rabbit
Immunogen A synthesized peptide derived from human PARD6B(Accession Q9BYG5), corresponding to amino acid residues T232-P282.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58220, 84612
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.