Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARD6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530854 Supplier Invitrogen™ Supplier No. PA530854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (85%), Pig (100%), Chicken (93%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain, an OPR domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cytoplasmic protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD6B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Pard6b
Gene Accession No. Q9BYG5
Gene Alias AV025615; par-6 (partitioning defective 6) homolog beta; PAR-6 beta; par-6 family cell polarity regulator beta; par-6 partitioning defective 6 homolog beta; par-6 partitioning defective 6 homolog beta (C. elegans); PAR6B; PAR-6B; Pard6b; pard6beta; partitioning defective 6 beta; partitioning defective 6 homolog beta; partitioning-defective protein 6, beta; si:dkey-192i18.6
Gene Symbols Pard6b
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PARD6B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.