Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARD6G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113332
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113332 Supplier Invitrogen™ Supplier No. PA5113332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARD6G Polyclonal Antibody for Western Blot, ICC/IF, ELISA

PAR6G involved in asymmetrical cell division and cell polarization processes. PAR6G may play a role in the formation of epithelial tight junctions. The PARD6-PARD3 complex links GTP-bound Rho small GTPases to atypical protein kinase C proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD6G
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PARD6G
Gene Accession No. Q9BYG4
Gene Alias 2410049N21Rik; par-6 family cell polarity regulator gamma; PAR-6 gamma; PAR-6 gamma protein; par-6 partitioning defective 6 homolog gamma; Par6a; PAR6D; PAR6G; PAR-6G; PAR6gamma; Pard6g; Partitioning defective 6 homolog gamma; partitioning-defective protein 6, alpha
Gene Symbols PARD6G
Host Species Rabbit
Immunogen Recombinant Human Partitioning defective 6 homolog gamma protein (249-326AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84552
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.