Learn More
Invitrogen™ PARD6G Polyclonal Antibody

Description
PARD6G Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PARD6G |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PARD6G |
| Gene Accession No. | Q9BYG4 |
| Gene Alias | 2410049N21Rik; par-6 family cell polarity regulator gamma; PAR-6 gamma; PAR-6 gamma protein; par-6 partitioning defective 6 homolog gamma; Par6a; PAR6D; PAR6G; PAR-6G; PAR6gamma; Pard6g; Partitioning defective 6 homolog gamma; partitioning-defective protein 6, alpha |
| Gene Symbols | PARD6G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.