Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARG Antibody (OTI6F4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273245AF488
Description
PARG Monoclonal specifically detects PARG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| PARG | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PARG | |
| Recombinant protein fragment corresponding to amino acids 1-367 of human PARG (NP_003622) produced in E.coli. | |
| 0.1 mL | |
| 8505.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6F4 | |
| Western Blot, Immunohistochemistry | |
| EC 3.2.1.143, FLJ54459, FLJ60257, FLJ60456, PARG99, poly (ADP-ribose) glycohydrolase, poly(ADP-ribose) glycohydrolase, poly(ADP-ribose) glycohydrolase 60 kDa isoform | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction