Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565755 Supplier Invitrogen™ Supplier No. PA565755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERDVVYFTFGD SELMRDIYSM HIFLTERKLT VGDVYKLLLR YYNEECRNCS TPGPDIKLYP FIYHAVESCA ETADHSGQRT GT Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PARG
Gene Accession No. Q86W56
Gene Alias AI413217; cytosolic poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase 63 kDa isoform; Parg; PARG99; poly (ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform
Gene Symbols PARG
Host Species Rabbit
Immunogen Recombinant Human PARG. Recombinant protein control fragment (Product #RP-106433).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8505
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.