Learn More
Invitrogen™ PARG Recombinant Rabbit Monoclonal Antibody (2B7M9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555512

Description
Immunogen Sequence: HNECLIITGT EQYSEYTGYA ETYRWSRSHE DGSERDDWQR RCTEIVAIDA LHFRRYLDQF VPEKMRRELN KAYCGFLRPG VSSENLSAVA TGNWGCGAFG GDARLKALIQ ILAAAAAERD VVYFTFGDSE LMRDIYSMHI FLTERKLTVG DVYKLLLRYY NEECRNCSTP GPDIKLYPFI YHAVESCAET ADHSGQRTGT.
Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.
Specifications
PARG | |
Recombinant Monoclonal | |
2.15 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O88622, Q86W56, Q9QYM2 | |
PARG | |
Recombinant Protein corresponding to a sequence within amino acids 777-976 of human PARG. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
2B7M9 | |
Unconjugated | |
PARG | |
AI413217; cytosolic poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase 63 kDa isoform; Parg; PARG99; poly (ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform | |
Rabbit | |
Protein A | |
RUO | |
26430, 83507, 8505 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.