Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARG1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105989DL594
Description
PARG1 Polyclonal antibody specifically detects PARG1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
PARG1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
PARG1PTPL1-associated RhoGAP 1 (PARG1), PTPL1-associated RhoGAP protein 1, Rho GTPase activating protein 29, rho GTPase-activating protein 29, Rho-type GTPase-activating protein 29, RP11-255E17.1 | |
The immunogen recognized by this antibody maps to a region between residue 1211 and 1261 of human PTPL1-associated RhoGAP 1 (NP_004806.3). | |
0.1 mL | |
Signal Transduction | |
9411 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction