Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555336 Supplier Invitrogen™ Supplier No. PA555336
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTTAPITISS LAEYSNQARL VEFLITYSQK IFDGSLQPQD VMCSIGVVDQ GCFPKPLLSP EERDIERSMK SLFFSSKEDI HTSESESKIF ERATSFEESE RKQNALGK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.

Rap1 is a small GTPase that, through effectors, regulates Rho GTPase signaling. These effectors- Rasip1, Radil, and the protein encoded by this gene- translocate to the cell membrane, where they form a multiprotein complex. This complex is necessary for Rap1-induced inhibition of Rho signaling. Defects in this gene may be a cause of nonsyndromic cleft lip with or without cleft palate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARG1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP29
Gene Accession No. Q52LW3
Gene Alias 6720461J18Rik; A830014I19; Arhgap29; AU040217; B130017I01Rik; C76601; Parg1; PTPL1-associated RhoGAP 1; PTPL1-associated RhoGAP 1 (PARG1); PTPL1-associated RhoGAP protein 1; RGD1306185; Rho GTPase activating protein 29; rho GTPase-activating protein 29; Rho-type GTPase-activating protein 29; RP11-255E17.1
Gene Symbols ARHGAP29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARG1. Recombinant protein control fragment (Product #RP-94245).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.