Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARG1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561331
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1331 Supplier Bethyl Laboratories, Inc Supplier No. A301923A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Rap1 is a small GTPase that, through effectors, regulates Rho GTPase signaling. These effectors- Rasip1, Radil, and the protein encoded by this gene- translocate to the cell membrane, where they form a multiprotein complex. This complex is necessary for Rap1-induced inhibition of Rho signaling. Defects in this gene may be a cause of nonsyndromic cleft lip with or without cleft palate.

Specifications

Antigen PARG1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ARHGAP29
Gene Accession No. Q52LW3
Gene Alias PARG1, RP11-255E17.1
Gene Symbols ARHGAP29
Host Species Rabbit
Immunogen Between 1125 and 1175
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9411
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.