Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Park7/DJ-1 Antibody (007), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP305805B
Description
Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
Park7/DJ-1 | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
007 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Park7/DJ-1 (Q99497-1; Met1-Asp189). | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
11315 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction