Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Park7/DJ-1 Antibody (4H4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$214.28 - $542.61
Specifications
| Antigen | Park7/DJ-1 |
|---|---|
| Clone | 4H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Simple Western 1:500, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP192715SS
![]() |
Novus Biologicals
NBP192715SS |
0.025 mL |
Each for $214.28
|
|
|||||
NBP192715
![]() |
Novus Biologicals
NBP192715 |
0.1 mL |
Each for $542.61
|
|
|||||
Description
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human, Bovine, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| Park7/DJ-1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| PARK7 | |
| IgG1 | |
| Protein G purified | |
| Human, Bovine and Mouse. |
| 4H4 | |
| Western Blot 1:5000, Simple Western 1:500, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Mouse (Negative), Rat (Negative) | |
| Q99497 | |
| 11315 | |
| Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title