Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARK9/ATP13A2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981601
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1601 Supplier Bioss Supplier No. BS11708R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neurodegenerative disorders such as Parkinson and Alzheimer disease cause motor and cognitive dysfunction and belong to a heterogeneous group of common and disabling disorders. ATP13A2, otherwise known as PARK9, is a neuronal P-type ATPase gene underlying an autosomal recessive form of early-onset parkinsonism with pyramidal degeneration and dementia. ATP13A2 protein is located in the membrane of these lysosomes and is formed most strongly in the brain, especially in the substantia nigra, a brain region which is known to play a central role in Parkinson′s disease.

Specifications

Antigen PARK9/ATP13A2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATP13A2
Gene Accession No. Q9NQ11
Gene Alias 1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4
Gene Symbols ATP13A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATP13A2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23400
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.