Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Parker™ ChromGas™ Zero Air Generator

Eliminates need for expensive gas cylinders

Supplier:  MilliporeSigma™ Supelco™ 27625U

 View more versions of this product

Catalog No. 11-100-2763


Description

Description

This ChromGas zero air generator incorporates three stages:

  • A 0.5 mm coalescing inlet filter removes particles, oil, and water
  • A heated catalyst removes hydrocarbons
  • A 0.01 mm cellulose fiber outlet filter removes residual particulate material from the product air stream
  • A hazard-free and economical alternative to high pressure gas cylinders, this unit should pay for itself in a year or two.

Features/Benefits

  • Eliminates need for expensive gas cylinders
  • Compact; requires less than one square foot of bench space
  • Can be used anywhere that an electrical supply is available
  • Only requires an upstream compressed air source

Specifications

  • Inlet Pressure: 2-125 psig
  • Outlet Purity: <0.1 ppm hydrocarbons (as methane)
  • Outlet Pressure: 125 psig
  • Outlet Fitting: 1/8 in. compression
  • Power Needs: 110 V (60 Hz), or 230 V (50 Hz)
  • Shipping Weight (Model 1000 and 1001): 11 lb (5 kg)
  • Shipping Weight (Model 3500 and 3501): 20 lb (9.1 kg)
  • Dimensions (Model 1000 and 1001; H x W x D): 9.75 in. x 5.75 in. x 12 in. (25 cm x 14.7 cm x 30.8 cm)
  • Dimensions (Model 3500 and 3501; H x W x D): 12 in. x 6.75 in. x 15 in. (29.2 cm x 17.8 cm x 39.4 cm)

Marks

  • CE approved
  • UL listed
  • CSA listed
  • IEC 1010 certified
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gas Chromatography
3500
110 Volt, Output Flow: 3,500 cc/min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

Parker is a registered trademark of Parker Hannifin