Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parkin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513420
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513420 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513420 Supplier Invitrogen™ Supplier No. PA513420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Parkin Polyclonal Antibody for Western Blot

The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parkin
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene park
Gene Accession No. Q7KTX7
Gene Alias AAN12155.1; AR-JP; CG10523; CG10523-PB; CG10523-PC; Dmel\CG10523; Dmel_CG10523; Dpark; dParkin; D-parkin; dpk; E3 ubiquitin ligase; E3 ubiquitin-protein ligase parkin; LPRS2; OTTHUMP00000017562; OTTHUMP00000017563; OTTHUMP00000017564; park; park1; PARK2; Parkin; parkin 2; parkin F; Parkin isoform 1; parkin protein; parkin RBR E3 ubiquitin protein ligase; Parkin RBR E3 ubiquitin-protein ligase; parkin variant SV5DEL; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin); park-PB; park-PC; PDJ; PK; Prkn; SD01679
Gene Symbols park
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 82-111 amino acids from the N-terminal region of human Drosophila Parkin.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40336
Target Species Drosophila
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.