Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parkin Recombinant Rabbit Monoclonal Antibody (21H24L9), Invitrogen™

Catalog No. 740019R
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-001-9R 200 μL
74-001-9R20UG 40 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-001-9R Supplier Invitrogen™ Supplier No. 740019R
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This is an engineered recombinant antibody that was developed to provide superior sensitivity without changing the paratope. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, lot-to-lot consistency and improved signal-to-noise ratio.

The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parkin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 21H24L9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRKN
Gene Accession No. O60260, Q9JK66, Q9WVS6
Gene Alias AAN12155.1; AR-JP; CG10523; CG10523-PB; CG10523-PC; Dmel\CG10523; Dmel_CG10523; Dpark; dParkin; D-parkin; dpk; E3 ubiquitin ligase; E3 ubiquitin-protein ligase parkin; LPRS2; OTTHUMP00000017562; OTTHUMP00000017563; OTTHUMP00000017564; park; park1; PARK2; Parkin; parkin 2; parkin F; Parkin isoform 1; parkin protein; parkin RBR E3 ubiquitin protein ligase; Parkin RBR E3 ubiquitin-protein ligase; parkin variant SV5DEL; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin); park-PB; park-PC; PDJ; PK; Prkn; SD01679
Gene Symbols PRKN
Host Species Rabbit
Immunogen Protein corresponding to human Parkin (aa1-aa465).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5071, 50873, 56816
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.