Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582683 Supplier Invitrogen™ Supplier No. PA582683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESLKSRVQSY FDGIKADWLD SIRPQKEGDF RKEINKWWNN LSDGQR Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PARL
Gene Accession No. Q9H300
Gene Alias D16Ertd607e; Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Parl; parl {ECO:0000250; Pbeta; P-beta; presenilin associated rhomboid like; presenilin associated, rhomboid-like; presenilin associated, rhomboid-like isoform 1 preproprotein; presenilin associated, rhomboid-like preproprotein; presenilins-associated rhomboid-like protein, mitochondrial; PRO2207; Psarl; PSARL1; PSENIP2; RHBDS1; rhomboid 7 homolog 1; UniProtKB:Q9H300}
Gene Symbols PARL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARL. Recombinant protein control fragment (Product #RP-91183).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55486
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.