Learn More
Invitrogen™ PARL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596978
Description
Immunogen sequence: MAWRGWAQRG WGCGQAWGAS VGGRSCEELT AVLTPPQLLG RRFNFFIQQK CGFRKAPRKV EPRRSDPGTS GEAYKRSALI PPVEETVFYP SPYPIRSLIK PLFFTVGFTG CAFGSAAIWQ YESLKSRVQS YFDGIKADWL DSIRPQKEGD FRKEINKWWN NLSDGQRTVT; Positive Samples: HepG2, MCF7; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein, Nucleus.
This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
PARL | |
Polyclonal | |
Unconjugated | |
PARL | |
D16Ertd607e; Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Parl; parl {ECO:0000250; Pbeta; P-beta; presenilin associated rhomboid like; presenilin associated, rhomboid-like; presenilin associated, rhomboid-like isoform 1 preproprotein; presenilin associated, rhomboid-like preproprotein; presenilins-associated rhomboid-like protein, mitochondrial; PRO2207; Psarl; PSARL1; PSENIP2; RHBDS1; rhomboid 7 homolog 1; UniProtKB:Q9H300} | |
Rabbit | |
Affinity chromatography | |
RUO | |
55486 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H300 | |
PARL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human PARL (NP_0010327281). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.