Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558307
Description
Immunogen sequence: VPPTTIWTSS PQNTDADTAS PSNGTHNNSV LPVTASAPTS LLPKNISIES REEEITSPGS NWEGTNTDPS PSGFSSTSGG VHLTTTLEEH SSGT Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 47%.
PARM1 may regulate TLP1 expression and telomerase activity, thus enabling certain prostatic cells to resist apoptosis.
Specifications
PARM1 | |
Polyclonal | |
Unconjugated | |
PARM1 | |
castration-induced prostatic apoptosis-related protein 1; Cipar1; DKFZP564O0823; PARM1; PARM-1; Prostate androgen-regulated mucin-like protein 1; prostatic androgen-regulated mucin-like protein 1; prostatic androgen-repressed message 1; prostatic androgen-repressed message-1; UNQ1879/PRO4322; WSC4; WSC4, cell wall integrity and stress response component 4 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWI2 | |
PARM1 | |
Recombinant protein corresponding to Human PARM1. Recombinant protein control fragment (Product #RP-96730). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction