Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARN Antibody (OTI4D12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273246F
Description
PARN Monoclonal specifically detects PARN in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
PARN | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PARN | |
Recombinant protein fragment corresponding to amino acids 1-266 of human PARN (NP_002573) produced in E.coli. | |
0.1 mL | |
5073.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4D12 | |
Immunohistochemistry | |
DANEC 3.1.13.4, Deadenylating nuclease, Deadenylation nuclease, poly(A)-specific ribonuclease (deadenylation nuclease), poly(A)-specific ribonuclease PARN, Polyadenylate-specific ribonuclease | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction