Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARN Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802721

Description
The protein encoded by this gene is a 3′-exoribonuclease, with similarity to the RNase D family of 3′-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
PARN | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95453 | |
PARN | |
Human recombit protein fragment corresponding to amino acids 1-266 of human PARN produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI2D9 | |
Unconjugated | |
PARN | |
DAN, DKCB6, PFBMFT4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5073 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction