Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530252 Supplier Invitrogen™ Supplier No. PA530252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NIH-3T3, BCL-1. Predicted reactivity: Mouse (87%), Rat (86%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a 3'-exoribonuclease, with similarity to the RNase D family of 3'-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARN
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Parn
Gene Accession No. O95453, Q8VDG3
Gene Alias 1200003I18Rik; DAN; Deadenylating nuclease; deadenylation nuclease; DKCB6; Parn; PFBMFT4; poly(A)-specific ribonuclease; poly(A)-specific ribonuclease (deadenylation nuclease); poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease; RGD1565449
Gene Symbols Parn
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PARN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5073, 74108
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.