Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Antibody (194C1439) - Cleaved - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 8 publications

$207.00 - $440.00

Specifications

Antigen PARP
Clone 194C1439
Concentration 0.5 mg/mL
Dilution Western Blot 1 - 2 ug/mL, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Flow (Intracellular) 1 - 10 ug/1x10^6 cells
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056599T
SDP
View Documents
Novus Biologicals
NB10056599SS
0.025 mg
Each for $207.00
Only null left
Add to Cart
 
NB10056599
SDP
View Documents
Novus Biologicals
NB10056599
0.1 mg
Each for $440.00
Only null left
Add to Cart
 
Description

Description

PARP Monoclonal specifically detects PARP in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications

Specifications

PARP
0.5 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Apoptosis, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Hypoxia, Necroptosis, Stem Cell Markers, Tumor Suppressors
P09874
142
This antibody was developed by immunizing mice with a synthetic peptide containing amino acids near 214/215-cleavage site of human PARP.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
194C1439
Western Blot 1 - 2 ug/mL, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Flow (Intracellular) 1 - 10 ug/1x10^6 cells
Monoclonal
Purified
RUO
Human, Mouse, Rat (Negative)
ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), cleaved PARP, cleaved PARP 1, cleaved PARP apoptosis, cleaved PARP western blot, cleaved PARP-1, EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1
PARP1
IgG2b κ
Protein G purified
This monoclonal antibody specifically detects a 89-kDa cleavage fragment containing the catalytic and auto modification domain.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.