Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PARP Inhibitor III, DPQ

Catalog No. 3002701mg Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg

The PARP Inhibitor III, DPQ, also referenced under CAS 129075-73-6, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

A potent and selective inhibitor of poly(ADP-ribose) polymerase (PARP; IC50 = 40 nM). Significantly reduces infarcet volume in rat focual cerebrale ischemia model.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Target Proteins PARP
CAS 129075-73-6
Molecular Formula C18H26N2O2
Formula Weight 302.4g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.