Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092444 20 μL
NB092443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092444 Supplier Novus Biologicals Supplier No. NBP30381720UL

Rabbit Polyclonal Antibody

PARP Polyclonal specifically detects PARP in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen PARP
Applications ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 81-390 of human PARP (NP_001609.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Apoptosis, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Hypoxia, Necroptosis, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 142
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.