Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP1 (cleaved Asp214) Recombinant Rabbit Monoclonal Antibody (PARP-H8), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$245.00 - $588.50
Specifications
Antigen | PARP1 (cleaved Asp214) |
---|---|
Clone | PARP-H8 |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537361
|
Invitrogen™
MA537361 |
10 Tests |
Each for $245.00
|
|
|||||
PIMA537115
![]() |
Invitrogen™
MA537115 |
100 Tests |
Each for $588.50
|
|
|||||
Description
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Specifications
PARP1 (cleaved Asp214) | |
Flow Cytometry | |
APC | |
Rabbit | |
Human | |
P09874 | |
142 | |
A synthetic peptide corresponding to residues surrounding Asp214 of human PARP. | |
Primary | |
4°C | |
PARP1 |
PARP-H8 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
5830444G22Rik; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+) poly (ADP-ribose) polymerase); ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase); ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1; ADP-ribosyltransferase NAD(+); Adprp; Adprt; ADPRT 1; ADPRT1; AI893648; ARTD1; C80510; DNA ADP-ribosyltransferase PARP1; msPARP; NAD(+) ADP-ribosyltransferase 1; pADPRT-1; PARP; Parp1; PARP-1; poly (ADP-ribose) polymerase 1; poly (ADP-ribose) polymerase family, member 1; poly (ADP-ribose) polymerase); poly [ADP-ribose] polymerase 1; poly(ADP-ribose) polymerase; poly(ADP-ribose) polymerase 1; poly(ADP-ribose) polymerase PARP-1; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase; poly[ADP-ribose] synthase 1; poly[ADP-ribose] synthetase 1; PPOL; Protein poly-ADP-ribosyltransferase PARP1; sPARP-1; synthetase fragment (AA 67); Unknown (protein for IMAGE:8023736); unnamed protein product | |
PARP1 | |
IgG κ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title