Learn More
PARP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | PARP1 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1814
![]() |
Bethyl Laboratories, Inc
IHC00279T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1813
![]() |
Bethyl Laboratories, Inc
IHC00279 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog, Horse, Orangutan, Monkey, Gorilla, Chimpanzee, Ferret and Crab-eating macaque
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Specifications
PARP1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PARP | |
PARP1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P09874 | |
142 | |
Between 50 and 100 | |
Primary | |
4° C | |
PARP1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.