Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP10 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002157FR
Description
PARP10 Polyclonal antibody specifically detects PARP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PARP10 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.4.2.30, FLJ14464, PARP-10, poly (ADP-ribose) polymerase family, member 10, poly [ADP-ribose] polymerase 10 | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of Poly (ADP-ribose) Polymerase Family, member 10 using the numbering given in entry NP_116178.1 (GeneID 84875) | |
0.1 mL | |
Apoptosis, DNA Repair, Hypoxia | |
84875 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction