Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555772 Supplier Invitrogen™ Supplier No. PA555772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHYDILEPEE LAENTSGGDH PSTQGPRATK HALLRTGGLV TALQGAGTVT MGSGEEPGQS GTSLRTGPMV QGRGIMTTGS GQEPGQSGTS L Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 46%.

Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors. Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors (Yu et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PARP10
Gene Accession No. Q53GL7
Gene Alias ADP-ribosyltransferase diphtheria toxin-like 10; ARTD10; FLJ14464; LOC102548778; Parp10; PARP-10; poly (ADP-ribose) polymerase family, member 10; poly [ADP-ribose] polymerase 10; poly [ADP-ribose] polymerase 10-like; poly(ADP-ribose) polymerase family member 10; Protein mono-ADP-ribosyltransferase PARP10
Gene Symbols PARP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARP10. Recombinant protein control fragment (Product #RP-93925).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.