Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP10 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | PARP10 |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5193
![]()
|
Proteintech Group Inc
260721AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.Specifications
| PARP10 | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q53GL7 | |
| 84875 | |
| PARP10 Fusion Protein Ag23457 | |
| Primary | |
| -20°C | |
| PARP10 |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| PARP 10, PARP10 | |
| PARP10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title