Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550474
Description
The antibody detects endogenous levels of total PARP11 protein.
Mono-ADP-ribosyltransferase that mediates mono-ADP-ribosylation of target proteins (PubMed:25043379, PubMed:25673562). Plays a role in nuclear envelope stability and nuclear remodeling during spermiogenesis. [UniProt].
Specifications
PARP11 | |
Polyclonal | |
Unconjugated | |
Parp11 | |
5330431N24Rik; ADP-ribosyltransferase diphtheria toxin-like 11; AI851877; ARTD11; C12orf6; HIN1L; MIB006; PARP11; PARP-11; poly (ADP-ribose) polymerase family, member 11; poly [ADP-ribose] polymerase 11; poly(ADP-ribose) polymerase family member 11; Protein mono-ADP-ribosyltransferase PARP11; RGD1561791 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57097 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NR21 | |
Parp11 | |
Full length fusion protein of Human PARP11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction