Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parp11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555456 Supplier Invitrogen™ Supplier No. PA555456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SISAFSYICE NEAIPMPPHW ENVNTQVPYQ LIPLHNQTHE YNEVANLFGK TMDRNRIKRI QRIQNLDLWE FFCRK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

Mono-ADP-ribosyltransferase that mediates mono-ADP-ribosylation of target proteins (PubMed:25043379, PubMed:25673562). Plays a role in nuclear envelope stability and nuclear remodeling during spermiogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parp11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Parp11
Gene Accession No. Q9NR21
Gene Alias 5330431N24Rik; ADP-ribosyltransferase diphtheria toxin-like 11; AI851877; ARTD11; C12orf6; HIN1L; MIB006; PARP11; PARP-11; poly (ADP-ribose) polymerase family, member 11; poly [ADP-ribose] polymerase 11; poly(ADP-ribose) polymerase family member 11; Protein mono-ADP-ribosyltransferase PARP11; RGD1561791
Gene Symbols Parp11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Parp11. Recombinant protein control fragment (Product #RP-89252).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.