Learn More
Invitrogen™ PARP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113581
Description
Immunogen sequence: IKHGNTFQIH GVSLQQRHLF RTYKSMFLAR VLIGDYINGD SKYMRPPSKD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Mono-ADP-ribosyltransferase that mediates mono-ADP-ribosylation of target proteins (PubMed:25043379, PubMed:25673562). Plays a role in nuclear envelope stability and nuclear remodeling during spermiogenesis. [UniProt].
Specifications
| PARP11 | |
| Polyclonal | |
| Unconjugated | |
| Parp11 | |
| 5330431N24Rik; ADP-ribosyltransferase diphtheria toxin-like 11; AI851877; ARTD11; C12orf6; HIN1L; MIB006; PARP11; PARP-11; poly (ADP-ribose) polymerase family, member 11; poly [ADP-ribose] polymerase 11; poly(ADP-ribose) polymerase family member 11; Protein mono-ADP-ribosyltransferase PARP11; RGD1561791 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NR21 | |
| Parp11 | |
| Synthetic peptide directed towards the middle region of human PARP11. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.