Learn More
Invitrogen™ PARP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589749
Description
Immunogen sequence: YVSPQDVTTM QTCNTKFPGP KSIPDYWDSS ALPDPGFQKI TLSSSSEEYQ KVWNLFNRTL PFYFVQKIER VQNLALWEVY QWQKGQMQKQ NGGKAVDERQ LFHGTSAIFV DAICQQNFDW RVCGVHGTSY GKGSYFARDA AYSHHYSKSD TQTHTMFLAR VLVGEFVRGN ASFVRPPAKE GWSNAFYDSC VNSVSDPSIF VIFEKHQVYP EYVIQYTTSS KPSVTPSILL ALGSLFSSRQ; Positive Samples: HT-29, HeLa, Mouse brain, Mouse liver, Mouse pancreas, Rat brain; Cellular Location: Nucleus.
Poly(ADP-ribosyl)ation is an immediate DNA-damage-dependent post-translational modification of histones and other nuclear proteins that contributes to the survival of injured proliferating cells. Poly(ADP-ribose) polymerases (PARPs) now constitute a large family of 18 proteins, encoded by different genes and displaying a conserved catalytic domain in which PARP-1 (113 kDa), the founding member, and PARP-2 (62 kDa) are so far the sole enzymes whose catalytic activity has been shown to be immediately stimulated by DNA strand breaks. A large repertoire of sequences encoding novel PARPs now extends considerably the field of poly(ADP-ribosyl)ation reactions to various aspects of the cell biology including cell proliferation and cell death. Some of these new members interact with each other, share common partners and common subcellular localizations suggesting possible fine tuning in the regulation of this post-translational modification of proteins.
Specifications
PARP12 | |
Polyclonal | |
Unconjugated | |
PARP12 | |
9930021O16; AA409132; AA536654; ADP-ribosyltransferase diphtheria toxin-like 12; ARTD12; MST109; MSTP109; PARP12; PARP-12; poly (ADP-ribose) polymerase family, member 12; poly [ADP-ribose] polymerase 12; poly ADP-ribose polymerase 12; poly(ADP-ribose) polymerase family member 12; Protein mono-ADP-ribosyltransferase PARP12; ZC3H1; Zc3hdc1; Zinc finger CCCH domain-containing protein 1; zinc finger CCCH type domain containing 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
243771, 362343, 64761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.92 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BZ20, Q9H0J9 | |
PARP12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 462-701 of human PARP12 (NP_0735871). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.