Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP14 Antibody (HL1178) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325591
Description
PARP14 Monoclonal antibody specifically detects PARP14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PARP14 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL1178 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:1000 | |
BAL2, KIAA1268, poly (ADP-ribose) polymerase family, member 14 | |
Recombinant protein encompassing a sequence within the C-terminus region of human PARP14. The exact sequence is proprietary. | |
100 μL | |
Apoptosis | |
54625 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction