Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540778 Supplier Invitrogen™ Supplier No. PA540778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: THGNHSLIVP PSKNPQNPTD LYDTVTDNVH HPSLFVAFYD YQAYPEYLIT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.

Poly [ADP-ribose] polymerase 14 (PARP14) is a B aggressive lymphoma protein belonging to the PARP protein family. Poly ADP ribosylation is a DNA damage dependent post translational modification of histones and other nuclear proteins that mediates DNA repair. PARP proteins with a macrodomain are also involved in gene expression through transcriptional regulation. PARP14 controls STAT6 dependent transcription by enhancing binding to the promoter in the presence of a positive signal (i.e. IL4 stimulation).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PARP14
Gene Accession No. Q460N5
Gene Alias 1600029O10Rik; ADP-ribosyltransferase diphtheria toxin-like 8; ARTD8; B aggressive lymphoma protein 2; B-aggressive lymphoma 2; BAL2; BC021340; CoaSt6; Collaborator of STAT6; KIAA1268; mKIAA1268; Parp14; PARP-14; pART8; poly (ADP-ribose) polymerase family, member 14; poly [ADP-ribose] polymerase 14; poly(ADP-ribose) polymerase family member 14; Protein mono-ADP-ribosyltransferase PARP14; protein mono-ADP-ribosyltransferase PARP14; poly [ADP-ribose] polymerase 14; RGD1310490; Unknown (protein for IMAGE:8227979)
Gene Symbols PARP14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PARP14 (aa 1749-1798).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.