Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP16 Mouse anti-Human, Clone: OTI4E2, liquid, TrueMAB™
SDP

Catalog No. 501671588
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1588 Supplier OriGene Supplier No. TA500453
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The function of this protein has not been specifically defined.

Specifications

Antigen PARP16
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PARP16
Gene Accession No. Q8N5Y8
Gene Alias ARTD15, C15orf30, pART15
Gene Symbols PARP16
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PARP16 expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54956
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.