Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP2 Antibody - Unpurified, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100185
Description
PARP2 Polyclonal specifically detects PARP2 in Human samples. It is validated for Western Blot.Specifications
PARP2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
PARP2 | |
Residues 43-59 [QRQESKKMPVAGGKANK] of the 62 kDa human PARP-2 protein. | |
Affinity Purified | |
RUO | |
Primary | |
N-terminus of human PARP 2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/mL | |
Western Blot 1:250-1:500 | |
ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2, ADPRT-2, ADPRT2pADPRT-2, ADPRTL2EC 2.4.2.30, ADPRTL3, hPARP-2, NAD(+) ADP-ribosyltransferase 2, PARP-2, poly (ADP-ribose) polymerase 2, poly (ADP-ribose) polymerase family, member 2, poly (ADP-ribosyl) transferase-like 2, poly [ADP-ribose] polymerase 2, poly(ADP-ribose) synthetase, Poly[ADP-ribose] synthase 2, poly[ADP-ribose] synthetase 2 | |
Rabbit | |
66 kDa | |
0.1 mL | |
Apoptosis, DNA Repair, Hypoxia | |
10038 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction