Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP2 Rabbit anti-Human, Mouse, Rat, Clone: 6G5U7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | PARP2 |
---|---|
Clone | 6G5U7 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PARP2 Monoclonal antibody specifically detects PARP2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PARP2 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2, ADPRT-2, ADPRT2pADPRT-2, ADPRTL2EC 2.4.2.30, ADPRTL3, hPARP-2, NAD(+) ADP-ribosyltransferase 2, PARP-2, poly (ADP-ribose) polymerase 2, poly (ADP-ribose) polymerase family, member 2, poly (ADP-ribosyl) transferase-like 2, poly [ADP-ribose] polymerase 2, poly(ADP-ribose) synthetase, Poly[ADP-ribose] synthase 2, poly[ADP-ribose] synthetase 2 | |
A synthetic peptide corresponding to a sequence within amino acids 484-583 of human PARP2 (Q9UGN5). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
6G5U7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, DNA Repair, Hypoxia | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10038 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title