Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | PARP3 |
---|---|
Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PARP3 Polyclonal antibody specifically detects PARP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PARP3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS (pH 7.2), 40% Glycerol | |
10039 | |
IgG | |
Immunogen affinity purified |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3, ADPRT-3, ADPRT3PADPRT-3, ADPRTL2, ADPRTL3NAD+ ADP-ribosyltransferase 3, EC 2.4.2.30, hPARP-3, IRT1ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2, NAD(+) ADP-ribosyltransferase 3, pADPRT-3, PARP-3, poly (ADP-ribose) polymerase family, member 3, poly [ADP-ribose] polymerase 3, poly(ADP-ribose) synthetase-3, Poly[ADP-ribose] synthase 3, poly[ADP-ribose] synthetase 3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LSKQQIARGFEALEALEEALKGPTDGGQSLEELSSHFYTVIPHNFGHSQPPPINSPELLQAKKDMLLVLADIELAQALQAVSEQEKTVEEV | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title