Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP3 Recombinant Rabbit Monoclonal Antibody (HL1509)
GREENER_CHOICE

Catalog No. PIMA547073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547073 Supplier Invitrogen™ Supplier No. MA547073
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1509
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PARP3
Gene Accession No. Q9Y6F1
Gene Alias A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; A930002C11Rik; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; Adprt3; ADPRT-3; ADPRTL2; Adprtl3; ARTD3; AW990611; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; Parp3; PARP-3; poly (ADP-ribose) polymerase family, member 3; poly (ADP-ribosyl) transferase-like 3; poly [ADP-ribose] polymerase 3; poly(ADP-ribose) polymerase family member 3; poly(ADP-ribose) synthetase-3; poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3
Gene Symbols PARP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PARP3. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10039, 235587
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.