Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584766 Supplier Invitrogen™ Supplier No. PA584766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSKQQIARGF EALEALEEAL KGPTDGGQSL EELSSHFYTV IPHNFGHSQP PPINSPELLQ AKKDMLLVLA DIELAQALQA VSEQEKTVEE V Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.

PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PARP3
Gene Accession No. Q9Y6F1
Gene Alias A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; A930002C11Rik; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; Adprt3; ADPRT-3; ADPRTL2; Adprtl3; ARTD3; AW990611; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; Parp3; PARP-3; poly (ADP-ribose) polymerase family, member 3; poly (ADP-ribosyl) transferase-like 3; poly [ADP-ribose] polymerase 3; poly(ADP-ribose) polymerase family member 3; poly(ADP-ribose) synthetase-3; poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3
Gene Symbols PARP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARP3. Recombinant protein control fragment (Product #RP-103952).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.