Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5139767

Catalog No. PIPA5139767


Only null left
Add to Cart

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AMAPKPKPWV QTEGPEKKKG RQAGREEDPF RSTAEALKAI PAEKRIIRVD PTCPLSSNPG TQVYEDYNCT LNQTNIENNN NKFYIIQLLQ DSNRFFTCW.

PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PARP3
Polyclonal
Unconjugated
PARP3
A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; A930002C11Rik; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; Adprt3; ADPRT-3; ADPRTL2; Adprtl3; ARTD3; AW990611; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; Parp3; PARP-3; poly (ADP-ribose) polymerase family, member 3; poly (ADP-ribosyl) transferase-like 3; poly [ADP-ribose] polymerase 3; poly(ADP-ribose) polymerase family member 3; poly(ADP-ribose) synthetase-3; poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3
Rabbit
Antigen affinity chromatography
RUO
10039
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y6F1
PARP3
Recombinant protein corresponding to Human PARP3. Recombinant protein control fragment (Product #RP-108741).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.