Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987737
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7737 Supplier Bioss Supplier No. BS7109R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.

Specifications

Antigen PARP4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PARP4
Gene Accession No. Q9UKK3
Gene Alias 193 kDa vault protein; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; ADPRTL1; ARTD4; C030027K23Rik; E230037B21Rik; Gm743; H5 proline-rich; I-alpha-I-related; Kiaa0177; p193; PARP4; PARP-4; PARPL; PARP-related; PARP-related/IalphaI-related H5/proline-rich; PH5P; poly (ADP-ribose) polymerase family, member 4; poly (ADP-ribose) polymerase)-like 1; Poly [ADP-ribose] polymerase 4; poly(ADP-ribose) polymerase family member 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; Protein mono-ADP-ribosyltransferase PARP4; Vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; VAULT3; von Willebrand factor A domain containing 5C; VPARP; VWA5C
Gene Symbols PARP4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PARP4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.